1-(5-Bromo-2-pyridinyl)-2-ethylpiperidine

Supplier: Matrix Scientific

052151-500MG 052151-2.5G
101898-494EA 364.62 USD
101898-494 101898-496
1-(5-Bromo-2-pyridinyl)-2-ethylpiperidine
1-(5-Bromo-2-pyridinyl)-2-ethylpiperidine

Hazard: Irritant

Formula: C₁₂H₁₇BrN₂
MDL Number: MFCD13561945
CAS Number: 1220030-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR