1-(5-Bromo-2-pyridinyl)-4-piperidinol

Supplier: Matrix Scientific

Synonyms: 1-(5-Bromopyridin-2-yl)piperidin-4-ol

052148-500MG 052148-2.5G
101898-482EA 538.46 USD
101898-482 101898-484
1-(5-Bromo-2-pyridinyl)-4-piperidinol
1-(5-Bromo-2-pyridinyl)-4-piperidinol

Hazard: Irritant

Formula: C₁₀H₁₃BrN₂O
MW: 257.14 g/mol
MDL Number: MFCD09910226
CAS Number: 149806-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR