4-(5-Bromo-2-pyridinyl)-2-piperazinone

Supplier: Matrix Scientific

052147-500MG 052147-2.5G
101898-478EA 364.62 USD
101898-478 101898-480
4-(5-Bromo-2-pyridinyl)-2-piperazinone
4-(5-Bromo-2-pyridinyl)-2-piperazinone

Hazard: Irritant

Formula: C₉H₁₀BrN₃O
MDL Number: MFCD13561943
CAS Number: 1197235-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR