1-(5-Bromo-2-pyridinyl)azepane

Supplier: Matrix Scientific

052146-500MG 052146-2.5G
101898-474EA 364.62 USD
101898-474 101898-476
1-(5-Bromo-2-pyridinyl)azepane
1-(5-Bromo-2-pyridinyl)azepane

Hazard: Irritant

Formula: C₁₁H₁₅BrN₂
MDL Number: MFCD11923073
CAS Number: 1175900-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR