1-(5-Bromo-2-pyridinyl)-4-methylpiperazine
Supplier: Matrix Scientific
Synonyms:
1-(5-Bromopyridin-2-yl)-4-methylpiperazine, 5-Bromo-2-(4-methylpiperazin-1-yl)pyridine
052144-500MG
052144-2.5G
101898-466EA
47.69
USD
101898-466
101898-468
1-(5-Bromo-2-pyridinyl)-4-methylpiperazine
1-(5-Bromo-2-pyridinyl)-4-methylpiperazine
Hazard: Irritant
Formula:
C₁₀H₁₄BrN₃ MW: 256.15 g/mol |
MDL Number:
MFCD07368225 CAS Number: 364794-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...