1-(5-Bromo-2-pyridinyl)-2-methylpiperidine

Supplier: Matrix Scientific

052141-500MG 052141-2.5G
101898-454EA 364.62 USD
101898-454 101898-456
1-(5-Bromo-2-pyridinyl)-2-methylpiperidine
1-(5-Bromo-2-pyridinyl)-2-methylpiperidine

Hazard: Irritant

Formula: C₁₁H₁₅BrN₂
MDL Number: MFCD13561940
CAS Number: 1220030-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR