2-[(5-Bromo-2-pyridinyl)(ethyl)amino]-1-ethanol

Supplier: Matrix Scientific

052139-500MG 052139-2.5G
101898-446EA 364.62 USD
101898-446 101898-448
2-[(5-Bromo-2-pyridinyl)(ethyl)amino]-1-ethanol
2-[(5-Bromo-2-pyridinyl)(ethyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₉H₁₃BrN₂O
MDL Number: MFCD13561938
CAS Number: 1220017-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR