5-Bromo-N-butyl-N-methyl-2-pyridinamine

Supplier: Matrix Scientific

052138-500MG 052138-2.5G
101898-442EA 364.62 USD
101898-442 101898-444
5-Bromo-N-butyl-N-methyl-2-pyridinamine
5-Bromo-N-butyl-N-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₀H₁₅BrN₂
MDL Number: MFCD13561937
CAS Number: 1220030-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR