1-(5-Bromo-2-pyridinyl)-3-pyrrolidinol

Supplier: Matrix Scientific

052137-500MG 052137-2.5G
101898-438EA 461.54 USD
101898-438 101898-440
1-(5-Bromo-2-pyridinyl)-3-pyrrolidinol
1-(5-Bromo-2-pyridinyl)-3-pyrrolidinol

Hazard: Irritant

Formula: C₉H₁₁BrN₂O
MDL Number: MFCD12033451
CAS Number: 1159816-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR