5-Bromo-2-(piperidin-1-yl)pyridine

Supplier: Matrix Scientific

052134-500MG 052134-2.5G
101898-426EA 364.62 USD
101898-426 101898-428
5-Bromo-2-(piperidin-1-yl)pyridine
5-Bromo-2-(piperidin-1-yl)pyridine

Hazard: Irritant

Formula: C₁₀H₁₃BrN₂
MDL Number: MFCD00234654
CAS Number: 24255-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR