5-Bromo-2-(1-pyrrolidinyl)pyridine

Supplier: Matrix Scientific

052132-500MG 052132-2.5G
101898-418EA 364.62 USD
101898-418 101898-420
5-Bromo-2-(1-pyrrolidinyl)pyridine
5-Bromo-2-(1-pyrrolidinyl)pyridine

Hazard: Irritant

Formula: C₉H₁₁BrN₂
MW: 227.11 g/mol
MDL Number: MFCD07781213
CAS Number: 210963-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR