2-[4-(5-Bromo-3-methyl-2-pyridinyl)-1-piperazinyl]-1-ethanol

Supplier: Matrix Scientific

052125-500MG 052125-2.5G
101898-390EA 364.62 USD
101898-390 101898-392
2-[4-(5-Bromo-3-methyl-2-pyridinyl)-1-piperazinyl]-1-ethanol
2-[4-(5-Bromo-3-methyl-2-pyridinyl)-1-piperazinyl]-1-ethanol

Hazard: Irritant

Formula: C₁₂H₁₈BrN₃O
MDL Number: MFCD13561931
CAS Number: 1220038-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR