N-Benzyl-5-bromo-N,3-dimethyl-2-pyridinamine

Supplier: Matrix Scientific

052123-500MG 052123-2.5G
101898-382EA 364.62 USD
101898-382 101898-384
N-Benzyl-5-bromo-N,3-dimethyl-2-pyridinamine
N-Benzyl-5-bromo-N,3-dimethyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₄H₁₅BrN₂
MDL Number: MFCD13561929
CAS Number: 446299-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR