N-(5-Bromo-3-methyl-2-pyridinyl)-N-cyclohexyl-N-methylamine

Supplier: Matrix Scientific

052121-500MG 052121-2.5G
101898-374EA 364.62 USD
101898-374 101898-376
N-(5-Bromo-3-methyl-2-pyridinyl)-N-cyclohexyl-N-methylamine
N-(5-Bromo-3-methyl-2-pyridinyl)-N-cyclohexyl-N-methylamine

Hazard: Irritant

Formula: C₁₃H₁₉BrN₂
MDL Number: MFCD13561927
CAS Number: 1220034-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR