5-Bromo-3-methyl-N,N-dipropyl-2-pyridinamine

Supplier: Matrix Scientific

052120-500MG 052120-2.5G
101898-370EA 364.62 USD
101898-370 101898-372
5-Bromo-3-methyl-N,N-dipropyl-2-pyridinamine
5-Bromo-3-methyl-N,N-dipropyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₉BrN₂
MDL Number: MFCD13561926
CAS Number: 1220036-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR