4-(5-Bromo-3-methyl-2-pyridinyl)-2-piperazinone

Supplier: Matrix Scientific

052118-500MG 052118-2.5G
101898-362EA 364.62 USD
101898-362 101898-364
4-(5-Bromo-3-methyl-2-pyridinyl)-2-piperazinone
4-(5-Bromo-3-methyl-2-pyridinyl)-2-piperazinone

Hazard: Irritant

Formula: C₁₀H₁₂BrN₃O
MDL Number: MFCD13561924
CAS Number: 1219968-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR