1-Benzyl-4-(5-bromo-3-methyl-2-pyridinyl)piperazine

Supplier: Matrix Scientific

052116-500MG 052116-2.5G
101898-354EA 364.62 USD
101898-354 101898-356
1-Benzyl-4-(5-bromo-3-methyl-2-pyridinyl)piperazine
1-Benzyl-4-(5-bromo-3-methyl-2-pyridinyl)piperazine

Hazard: Irritant

Formula: C₁₇H₂₀BrN₃
MDL Number: MFCD13561922
CAS Number: 1220030-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR