1-(5-Bromo-3-methyl-2-pyridinyl)-4-methylpiperazine

Supplier: Matrix Scientific

052115-500MG 052115-2.5G
101898-350EA 364.62 USD
101898-350 101898-352
1-(5-Bromo-3-methyl-2-pyridinyl)-4-methylpiperazine
1-(5-Bromo-3-methyl-2-pyridinyl)-4-methylpiperazine

Hazard: Irritant

Formula: C₁₁H₁₆BrN₃
MDL Number: MFCD13561921
CAS Number: 885267-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR