5-Bromo-3-methyl-2-(2-methyl-1-piperidinyl)pyridine

Supplier: Matrix Scientific

052112-500MG 052112-2.5G
101898-338EA 364.62 USD
101898-338 101898-340
5-Bromo-3-methyl-2-(2-methyl-1-piperidinyl)pyridine
5-Bromo-3-methyl-2-(2-methyl-1-piperidinyl)pyridine

Hazard: Irritant

Formula: C₁₂H₁₇BrN₂
MDL Number: MFCD13561918
CAS Number: 1220036-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR