N,N-Diallyl-5-bromo-3-methyl-2-pyridinamine

Supplier: Matrix Scientific

052111-500MG 052111-2.5G
101898-334EA 364.62 USD
101898-334 101898-336
N,N-Diallyl-5-bromo-3-methyl-2-pyridinamine
N,N-Diallyl-5-bromo-3-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₅BrN₂
MDL Number: MFCD13561917
CAS Number: 1220036-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR