2-[(5-Bromo-3-methyl-2-pyridinyl)(ethyl)amino]-1-ethanol

Supplier: Matrix Scientific

052110-500MG 052110-2.5G
101898-330EA 364.62 USD
101898-330 101898-332
2-[(5-Bromo-3-methyl-2-pyridinyl)(ethyl)amino]-1-ethanol
2-[(5-Bromo-3-methyl-2-pyridinyl)(ethyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₁₀H₁₅BrN₂O
MDL Number: MFCD13561916
CAS Number: 1219967-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR