5-Bromo-N-butyl-N,3-dimethyl-2-pyridinamine

Supplier: Matrix Scientific

052109-500MG 052109-2.5G
101898-326EA 364.62 USD
101898-326 101898-328
5-Bromo-N-butyl-N,3-dimethyl-2-pyridinamine
5-Bromo-N-butyl-N,3-dimethyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₁H₁₇BrN₂
MDL Number: MFCD13561915
CAS Number: 1220036-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR