1-(5-Bromo-3-methyl-2-pyridinyl)-3-pyrrolidinol

Supplier: Matrix Scientific

052108-500MG 052108-2.5G
101898-322EA 364.62 USD
101898-322 101898-324
1-(5-Bromo-3-methyl-2-pyridinyl)-3-pyrrolidinol
1-(5-Bromo-3-methyl-2-pyridinyl)-3-pyrrolidinol

Hazard: Irritant

Formula: C₁₀H₁₃BrN₂O
MW: 257.14 g/mol
MDL Number: MFCD13561914
CAS Number: 1220016-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR