2-[(5-Bromo-3-methyl-2-pyridinyl)(methyl)amino]-1-ethanol

Supplier: Matrix Scientific

052105-500MG 052105-2.5G
101898-310EA 364.62 USD
101898-310 101898-312
2-[(5-Bromo-3-methyl-2-pyridinyl)(methyl)amino]-1-ethanol
2-[(5-Bromo-3-methyl-2-pyridinyl)(methyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₉H₁₃BrN₂O
MDL Number: MFCD13561912
CAS Number: 1220034-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR