5-Bromo-N,N-diethyl-3-methyl-2-pyridinamine

Supplier: Matrix Scientific

052104-500MG 052104-2.5G
101898-306EA 364.62 USD
101898-306 101898-308
5-Bromo-N,N-diethyl-3-methyl-2-pyridinamine
5-Bromo-N,N-diethyl-3-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₀H₁₅BrN₂
MDL Number: MFCD13561911
CAS Number: 1220038-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR