5-Bromo-N,N,3-trimethyl-2-pyridinamine

Supplier: Matrix Scientific

052102-500MG 052102-2.5G
101898-298EA 461.54 USD
101898-298 101898-300
5-Bromo-N,N,3-trimethyl-2-pyridinamine
5-Bromo-N,N,3-trimethyl-2-pyridinamine

Hazard: Irritant

Formula: C₈H₁₁BrN₂
MDL Number: MFCD13561909
CAS Number: 245765-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR