N2-Benzyl-6-chloro-N2-ethyl-1,3,5-triazine-2,4-diamine

Supplier: Matrix Scientific

052100-500MG 052100-2.5G
101898-290EA 364.62 USD
101898-290 101898-292
N2-Benzyl-6-chloro-N2-ethyl-1,3,5-triazine-2,4-diamine
N2-Benzyl-6-chloro-N2-ethyl-1,3,5-triazine-2,4-diamine

Hazard: Irritant

Formula: C₁₂H₁₄ClN₅
MDL Number: MFCD13561907
CAS Number: 1220029-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR