2-[4-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-1-piperazinyl]-1-ethanol

Supplier: Matrix Scientific

052096-500MG 052096-2.5G
101898-274EA 364.62 USD
101898-274 101898-276
2-[4-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-1-piperazinyl]-1-ethanol
2-[4-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-1-piperazinyl]-1-ethanol

Hazard: Irritant

Formula: C₉H₁₅ClN₆O
MDL Number: MFCD13561903
CAS Number: 1220016-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR