N-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-N-cyclohexyl-N-ethylamine

Supplier: Matrix Scientific

052095-500MG 052095-2.5G
101898-270EA 364.62 USD
101898-270 101898-272
N-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-N-cyclohexyl-N-ethylamine
N-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-N-cyclohexyl-N-ethylamine

Hazard: Irritant

Formula: C₁₁H₁₈ClN₅
MDL Number: MFCD13561902
CAS Number: 1219967-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR