N-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-N-cyclohexyl-N-methylamine

Supplier: Matrix Scientific

052092-500MG 052092-2.5G
101898-258EA 364.62 USD
101898-258 101898-260
N-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-N-cyclohexyl-N-methylamine
N-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-N-cyclohexyl-N-methylamine

Hazard: Irritant

Formula: C₁₀H₁₆ClN₅
MDL Number: MFCD13561899
CAS Number: 99192-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR