1-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-4-piperidinol

Supplier: Matrix Scientific

052090-500MG 052090-2.5G
101898-250EA 364.62 USD
101898-250 101898-252
1-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-4-piperidinol
1-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-4-piperidinol

Hazard: Irritant

Formula: C₈H₁₂ClN₅O
MDL Number: MFCD13561898
CAS Number: 1220016-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR