4-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-2-piperazinone

Supplier: Matrix Scientific

052089-500MG 052089-2.5G
101898-246EA 364.62 USD
101898-246 101898-248
4-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-2-piperazinone
4-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-2-piperazinone

Hazard: Irritant

Formula: C₇H₉ClN₆O
MDL Number: MFCD13561897
CAS Number: 1220029-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR