4-Chloro-6-(3-methyl-1-piperidinyl)-1,3,5-triazin-2-amine

Supplier: Matrix Scientific

052084-500MG 052084-2.5G
101898-226EA 364.62 USD
101898-226 101898-228
4-Chloro-6-(3-methyl-1-piperidinyl)-1,3,5-triazin-2-amine
4-Chloro-6-(3-methyl-1-piperidinyl)-1,3,5-triazin-2-amine

Hazard: Irritant

Formula: C₉H₁₄ClN₅
MDL Number: MFCD13561892
CAS Number: 1220033-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR