2-[(4-Amino-6-chloro-1,3,5-triazin-2-yl)(ethyl)amino]-1-ethanol

Supplier: Matrix Scientific

052081-500MG 052081-2.5G
101898-214EA 364.62 USD
101898-214 101898-216
2-[(4-Amino-6-chloro-1,3,5-triazin-2-yl)(ethyl)amino]-1-ethanol
2-[(4-Amino-6-chloro-1,3,5-triazin-2-yl)(ethyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₇H₁₂ClN₅O
MDL Number: MFCD13561889
CAS Number: 856369-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR