1-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-3-pyrrolidinol

Supplier: Matrix Scientific

052079-500MG 052079-2.5G
101898-206EA 364.62 USD
101898-206 101898-208
1-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-3-pyrrolidinol
1-(4-Amino-6-chloro-1,3,5-triazin-2-yl)-3-pyrrolidinol

Hazard: Irritant

Formula: C₇H₁₀ClN₅O
MDL Number: MFCD13561887
CAS Number: 1220030-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR