2-[(4-Amino-6-chloro-1,3,5-triazin-2-yl)(methyl)amino]-1-ethanol

Supplier: Matrix Scientific

052076-500MG 052076-2.5G
101898-194EA 364.62 USD
101898-194 101898-196
2-[(4-Amino-6-chloro-1,3,5-triazin-2-yl)(methyl)amino]-1-ethanol
2-[(4-Amino-6-chloro-1,3,5-triazin-2-yl)(methyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₆H₁₀ClN₅O
MDL Number: MFCD13561886
CAS Number: 1220031-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR