{6-[3,4-Dihydro-1(2H)-quinolinyl]-3-pyridinyl}methanamine

Supplier: Matrix Scientific

052071-500MG 052071-2.5G
101898-174EA 364.62 USD
101898-174 101898-176
{6-[3,4-Dihydro-1(2H)-quinolinyl]-3-pyridinyl}methanamine
{6-[3,4-Dihydro-1(2H)-quinolinyl]-3-pyridinyl}methanamine

Hazard: Irritant

Formula: C₁₅H₁₇N₃
MDL Number: MFCD09741881
CAS Number: 954567-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR