[6-(2,3-Dihydro-1H-indol-1-yl)-3-pyridinyl]methanamine

Supplier: Matrix Scientific

052070-500MG 052070-2.5G
101898-170EA 364.62 USD
101898-170 101898-172
[6-(2,3-Dihydro-1H-indol-1-yl)-3-pyridinyl]methanamine
[6-(2,3-Dihydro-1H-indol-1-yl)-3-pyridinyl]methanamine

Hazard: Irritant

Formula: C₁₄H₁₅N₃
MDL Number: MFCD09734258
CAS Number: 953726-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR