{6-[3,4-Dihydro-2(1H)-isoquinolinyl]-3-pyridinyl}methanamine

Supplier: Matrix Scientific

052069-500MG 052069-2.5G
101898-166EA 364.62 USD
101898-166 101898-168
{6-[3,4-Dihydro-2(1H)-isoquinolinyl]-3-pyridinyl}methanamine
{6-[3,4-Dihydro-2(1H)-isoquinolinyl]-3-pyridinyl}methanamine

Hazard: Irritant

Formula: C₁₅H₁₇N₃
MDL Number: MFCD09739070
CAS Number: 954269-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR