5-(Aminomethyl)-N-cyclohexyl-N-ethyl-2-pyridinamine

Supplier: Matrix Scientific

052067-500MG 052067-2.5G
101898-158EA 364.62 USD
101898-158 101898-160
5-(Aminomethyl)-N-cyclohexyl-N-ethyl-2-pyridinamine
5-(Aminomethyl)-N-cyclohexyl-N-ethyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₄H₂₃N₃
MDL Number: MFCD09737194
CAS Number: 954273-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR