[6-(2-Ethyl-1-piperidinyl)-3-pyridinyl]methanamine

Supplier: Matrix Scientific

052065-500MG 052065-2.5G
101898-150EA 364.62 USD
101898-150 101898-152
[6-(2-Ethyl-1-piperidinyl)-3-pyridinyl]methanamine
[6-(2-Ethyl-1-piperidinyl)-3-pyridinyl]methanamine

Hazard: Irritant

Formula: C₁₃H₂₁N₃
MDL Number: MFCD11624785
CAS Number: 1094798-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR