5-(Aminomethyl)-N-cyclohexyl-N-methyl-2-pyridinamine

Supplier: Matrix Scientific

052064-500MG 052064-2.5G
101898-146EA 364.62 USD
101898-146 101898-148
5-(Aminomethyl)-N-cyclohexyl-N-methyl-2-pyridinamine
5-(Aminomethyl)-N-cyclohexyl-N-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₃H₂₁N₃
MDL Number: MFCD09727826
CAS Number: 953719-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR