5-(Aminomethyl)-N,N-dipropyl-2-pyridinamine

Supplier: Matrix Scientific

052063-500MG 052063-2.5G
101898-142EA 186.15 USD
101898-142 101898-144
5-(Aminomethyl)-N,N-dipropyl-2-pyridinamine
5-(Aminomethyl)-N,N-dipropyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₂₁N₃
MDL Number: MFCD09044404
CAS Number: 926218-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR