1-[5-(Aminomethyl)-2-pyridinyl]-4-piperidinol

Supplier: Matrix Scientific

052062-500MG 052062-2.5G
101898-138EA 186.15 USD
101898-138 101898-140
1-[5-(Aminomethyl)-2-pyridinyl]-4-piperidinol
1-[5-(Aminomethyl)-2-pyridinyl]-4-piperidinol

Hazard: Irritant

Formula: C₁₁H₁₇N₃O
MDL Number: MFCD09728442
CAS Number: 953751-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR