4-[5-(Aminomethyl)-2-pyridinyl]-2-piperazinone

Supplier: Matrix Scientific

052061-500MG 052061-2.5G
101898-134EA 384.62 USD
101898-134 101898-136
4-[5-(Aminomethyl)-2-pyridinyl]-2-piperazinone
4-[5-(Aminomethyl)-2-pyridinyl]-2-piperazinone

Hazard: Irritant

Formula: C₁₀H₁₄N₄O
MDL Number: MFCD09050451
CAS Number: 926245-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR