[6-(1-Azepanyl)-3-pyridinyl]methanamine

Supplier: Matrix Scientific

Synonyms: (6-(Azepan-1-yl)pyridin-3-yl)methanamine

052060-500MG 052060-2.5G
101898-130EA 364.62 USD
101898-130 101898-132
[6-(1-Azepanyl)-3-pyridinyl]methanamine
[6-(1-Azepanyl)-3-pyridinyl]methanamine

Hazard: Irritant

Formula: C₁₂H₁₉N₃
MW: 205.31 g/mol
MDL Number: MFCD09045371
CAS Number: 926204-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR