[6-(4-Benzyl-1-piperazinyl)-3-pyridinyl]methanamine

Supplier: Matrix Scientific

052059-500MG 052059-2.5G
101898-126EA 364.62 USD
101898-126 101898-128
[6-(4-Benzyl-1-piperazinyl)-3-pyridinyl]methanamine
[6-(4-Benzyl-1-piperazinyl)-3-pyridinyl]methanamine

Hazard: Irritant

Formula: C₁₇H₂₂N₄
MDL Number: MFCD13561884
CAS Number: 1156349-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR