[6-(4-Methyl-1-piperazinyl)-3-pyridinyl]methanamine

Supplier: Matrix Scientific

052058-500MG 052058-2.5G
101898-122EA 461.54 USD
101898-122 101898-124
[6-(4-Methyl-1-piperazinyl)-3-pyridinyl]methanamine
[6-(4-Methyl-1-piperazinyl)-3-pyridinyl]methanamine

Hazard: Irritant

Formula: C₁₁H₁₈N₄
MDL Number: MFCD08729232
CAS Number: 767628-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR