[6-(4-Methyl-1-piperidinyl)-3-pyridinyl]methanamine

Supplier: Matrix Scientific

052057-500MG 052057-2.5G
101898-118EA 364.62 USD
101898-118 101898-120
[6-(4-Methyl-1-piperidinyl)-3-pyridinyl]methanamine
[6-(4-Methyl-1-piperidinyl)-3-pyridinyl]methanamine

Hazard: Irritant

Formula: C₁₂H₁₉N₃
MDL Number: MFCD09044322
CAS Number: 926266-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR