[6-(3-Methyl-1-piperidinyl)-3-pyridinyl]methanamine

Supplier: Matrix Scientific

052056-500MG 052056-2.5G
101898-114EA 364.62 USD
101898-114 101898-116
[6-(3-Methyl-1-piperidinyl)-3-pyridinyl]methanamine
[6-(3-Methyl-1-piperidinyl)-3-pyridinyl]methanamine

Hazard: Irritant

Formula: C₁₂H₁₉N₃
MDL Number: MFCD09043008
CAS Number: 926196-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR